Not web orderable
The product 02681440: SureOne™ 10uL Ultra Micropoint Pipette Tip, Universal Fit is not orderable in your country at the moment. Please contact our Technical Support for possible alternatives.
Explore other products in this category
Universal Pipette Tips
Universal pipette tips are disposable and designed to fit multichannel and single pipettes from many manufacturers. Compared to pipette specific tips, universal pipette tips offer high levels of performance and offer versatile uses with many pipette models. Thermo Scientific™ ART™ Barrier Reload Insert Pipette Tips seal when exposed to potential contaminants to provide consistent and accurate performance.