Thermo Scientific Chemicals

N-Octadecylphosphonsäure, 97 %, Thermo Scientific Chemicals

Katalognummer: 020645.06
5 g, Each
Thermo Scientific Chemicals

N-Octadecylphosphonsäure, 97 %, Thermo Scientific Chemicals

Katalognummer: 020645.06
5 g, Each
Menge
Katalognummer: 020645.06
auch als 020645-06 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
4724-47-4
IUPAC Name
octadecylphosphonic acid
Molecular Formula
C18H39O3P
InChI Key
FTMKAMVLFVRZQX-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCP(O)(O)=O
Appearance (Color)
White to pale cream
Assay (GC)
≥96.0%
Melting Point (DSC)
95 deg C, min

Beschreibung

n-Octadecylphosphonic acid is used in thermal paper for receipts, adding machines and tickets. It acts as surfactant, emulsifier, dispersants and chelating agents. Further, it is a long chain phosphonic acid compound utilized for proteomics research. In addition to this, it is used to prepare modified mesoporous magnetic nano particles through Lewis acid/base interaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P332 + P313 - If skin irritation occurs: Get medical advice/attention

    P337 + P313 - If eye irritation persists: Get medical advice/attention