Gallium(III)-bromid, wasserfrei, Thermo Scientific Chemicals
Gallium(III)-bromid, wasserfrei, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gallium(III)-bromid, wasserfrei, Thermo Scientific Chemicals

Have Questions?
Menge:
10 g
Katalognummer 032107.09
auch als 032107-09 bezeichnet
Preis (EUR)/ Each
329,40
Exklusiv online
366.00 
Ersparnis 36,60 (10%)
-
Zum Warenkorb hinzufügen
Menge:
10 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)/ Each
329,40
Exklusiv online
366.00 
Ersparnis 36,60 (10%)
Zum Warenkorb hinzufügen
Gallium(III)-bromid, wasserfrei, Thermo Scientific Chemicals
Katalognummer032107.09
Preis (EUR)/ Each
329,40
Exklusiv online
366.00 
Ersparnis 36,60 (10%)
-
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS13450-88-9
IUPAC Namegallium(3+) tribromide
Molecular FormulaBr3Ga
InChI KeySRVXDMYFQIODQI-UHFFFAOYSA-K
SMILES[Ga+3].[Br-].[Br-].[Br-]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Assay (unspecified)99% min
Gallium(III) bromide is used as organic intermediates. It is a powerful oxidizing agent and is able to release oxygen free radicals from the water in mucous membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only