Platin-Iridium-Draht, 1.0 mm (0.04 in.) Durchmesser, hart, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platin-Iridium-Draht, 1.0 mm (0.04 in.) Durchmesser, hart, Thermo Scientific Chemicals
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
10 cm
5 cm
50 cm
Katalognummer 044732.BS
auch als 044732-BS bezeichnet
Preis (EUR)/ Each
-
Menge:
10 cm
Großbestellung oder individuelle Größe anfordern
SpecificationsSpecification SheetDatenblatt
FormWire, 1.0mm (0.04in) diameter, hardened.
Appearance (Color)Silver gray
Assay from Supplier's CofAPt:Ir; 80:20 wt%
Platinum Iridium wire, 1.0mm (0.04in) dia, hard is used in non-standard thermocouples, electrical contacts and as electrodes in medical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only