MDS Mobile Detection Systems
MDS Mobile Detection Systems
Thermo Scientific™

MDS Mobile Detection Systems

The Thermo Scientific™ MDS Mobile Detection System is a mobile radiation reconnaissance system designed to operate in a vehicle for use on the ground or in airspace.
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerBeschreibung
4255004Mobile radiation detection system
4255009Gamma radiation reconnaissance
Katalognummer 4255004
Preis (EUR)
-
Angebot anfordern
Beschreibung:
Mobile radiation detection system
Ask our AI about this Product
The Thermo Scientific™ MDS Mobile Detection System is a mobile radiation reconnaissance system designed to operate in a vehicle for use on the ground or in airspace. It is for use around or in airspace to discover/map the location of artificial radiation sources or survey large areas of contamination.
  • Rugged, easily relocated military specification system with user friendly features
  • Allows for a wide range of users without compromising accuracy and results
  • Remote viewing and/or control of mission from command station via wireless transmission of data
  • Fully integrated ArcView GIS for simultaneous hazard modeling and verification with real time data
  • Fleet management capabilities allows for use of many detectors and/or personnel and asset location
  • Lightweight, modular design allows for easy relocation between vehicles
  • No hard wiring into vehicles is required
  • Real time data feed into any version
  • ESRI ArcView software is standard
  • Optional neutron and contamination probes to detect and measure all types of radiation
  • NBR technology for instant threat assessment of radiological incidents
  • Data transmission by radio modem or GSM wireless devices
Ordering Alerts:
  • Optional Neutron Detector is He3 detector filled to 10 atmospheres
Includes:
  • Zargus military grade carrying case, NATO approved
Recommended for:
  • Tactical hazmat operations for civilian and military responders (CST's, police, fire, hazmat and radiological response teams)
  • Surveillance and intelligence gathering
  • Geo spatial intelligence analysis
  • Special event monitoring
  • Aerial surveys of contaminated areas
Specifications
BeschreibungMobile radiation detection system
DetektortypScintillation Detector
Höhe (angloamerikanisch)13.8 in.
Höhe (metrisch)35 cm
Tiefe (angloamerikanisch)15.7 in.
Tiefe (metrisch)40 cm
MessbereichScintillation Detector: 1 nSv/h to 20 µSv/h; Proportional Detector: 10 nSv/h to 1 Sv/h
StrahlungstypDependent on probe used
Empfindlichkeit2μSv/h (0.2μR/h) increase above background
Gewicht (angloamerikanisch)33.1 lb.
Gewicht (metrisch)15 kg
Breite (angloamerikanisch)23.6 in.
Breite (metrisch)60 cm
Relative Luftfeuchtigkeit5% to 95% Non-Condensing
Temperaturbereich (angloamerikanisch)32°F to 122°F
Temperaturbereich (metrisch)0°C to 50°C
Schwellengrenzwert50 keV
ProdukttypMobile Detection System
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Dokumente und Downloads

Zertifikate

0 Ergebnisse angezeigt, oben nach einem bestimmten Zertifikat suchen

Sicherheitsdatenblätter

Geben Sie die Katalognummer, den Namen oder Link frei.

1x1 image pixel for data collection