Triamcinolon
Triamcinolon
Triamcinolon
Thermo Scientific Chemicals

Triamcinolon

Have Questions?
KatalognummerMenge
4622400505g
Katalognummer 462240050
Preis (EUR)
257,55
Exklusiv online
311,00
Ersparnis 53,45 (17%)
Each
-
Zum Warenkorb hinzufügen
Menge:
5g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
257,55
Exklusiv online
311,00
Ersparnis 53,45 (17%)
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS124-94-7
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White to off-white
Loss on drying=<2 %
HPLC>=96.0 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Dokumente und Downloads

Zertifikate

Sicherheitsdatenblätter

Sicherheit und Handhabung

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Carcinogenicity
Category 2
Label Elements
Signal Word

Warning

Hazard Statements

H351 - Suspected of causing cancer

Precautionary Statements

P201 - Obtain special instructions before use

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P308 + P313 - IF exposed or concerned: Get medical advice/attention

Geben Sie die Katalognummer, den Namen oder Link frei.