Bambuterol-Hydrochlorid, Thermo Scientific Chemicals
Product Image
Thermo Scientific Chemicals

Bambuterol-Hydrochlorid, Thermo Scientific Chemicals

Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerMenge
4649100505 g
4649100101 g
Katalognummer 464910050
Preis (EUR)
232,90
Online Exclusive
274,00
Ersparnis 41,10 (15%)
Each
Zum Warenkorb hinzufügen
Menge:
5 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
232,90
Online Exclusive
274,00
Ersparnis 41,10 (15%)
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS81732-46-9
IUPAC Namehydrogen 3-[2-(tert-butylamino)-1-hydroxyethyl]-5-[(dimethylcarbamoyl)oxy]phenyl N,N-dimethylcarbamate chloride
Molecular FormulaC18H30ClN3O5
InChI KeyLBARATORRVNNQM-UHFFFAOYNA-N
SMILES[H+].[Cl-].CN(C)C(=O)OC1=CC(=CC(OC(=O)N(C)C)=C1)C(O)CNC(C)(C)C
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Infrared spectrumConforms
Appearance (Color)White to light yellow
Water=<0.5 %
Titration Argentometric>=97.5 %
Appearance (Form)Crystalline powder
Mehr anzeigen
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Dokumente und Downloads

Zertifikate

Chargen-Nr.Certificate TypeDateCatalog Number(s)
A0414567Certificate of Analysis25. Jan. 2025464910050, 464910010
A0414567Certificate of Analysis25. Juli 2022464910050, 464910010
2 Ergebnisse angezeigt, oben nach einem bestimmten Zertifikat suchen

Sicherheitsdatenblätter

Geben Sie die Katalognummer, den Namen oder Link frei.

1x1 image pixel for data collection