1x1 image pixel for data collection
Flunixin-Meglumin
Flunixin-Meglumin
Thermo Scientific Chemicals

Flunixin-Meglumin

Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerMenge
467162500250 mg
4671600101 g
Katalognummer 467162500
Preis (EUR)
112,20
Online Exclusive
135,00
Ersparnis 22,80 (17%)
Each
Zum Warenkorb hinzufügen
Menge:
250 mg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
112,20
Online Exclusive
135,00
Ersparnis 22,80 (17%)
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS42461-84-7
IUPAC Name(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol; 2-{[2-methyl-3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid
Molecular FormulaC21H28F3N3O7
InChI KeyMGCCHNLNRBULBU-WZTVWXICSA-N
SMILESCNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.CC1=C(NC2=NC=CC=C2C(O)=O)C=CC=C1C(F)(F)F
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Titration with HClO498.0 to 102.0 %
Appearance (Color)White to beige
Appearance (Form)Powder
Infrared spectrumConforms
Melting point136°C to 138°C
Mehr anzeigen
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Geben Sie die Katalognummer, den Namen oder Link frei.