Thermo Scientific Chemicals

Chlortetracycline hydrochloride, Thermo Scientific Chemicals

Katalognummer: J60095.22
100 g, Each
Thermo Scientific Chemicals

Chlortetracycline hydrochloride, Thermo Scientific Chemicals

Katalognummer: J60095.22
100 g, Each
Menge
Informational:Informatorisch:Für diese Chemikalie benötigen wir möglicherweise zusätzliche Informationen für unsere Unterlagen zur behördlichen und chemischen Konformität. Falls erforderlich, setzen wir uns mit Ihnen in Verbindung, sobald Ihre Bestellung eingegangen ist, um diese Informationen zu erfragen.
Katalognummer: J60095.22
auch als J60095-22 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
64-72-2
IUPAC Name
hydrogen (2Z,4S,4aS,5aS,6S,12aS)-2-[amino(hydroxy)methylidene]-7-chloro-4-(dimethylamino)-6,10,11,12a-tetrahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
Molecular Formula
C22H24Cl2N2O8
InChI Key
RRXOSDMYCQSQHB-MRYPCDLHSA-N
SMILES
[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)C4=C(O)C=CC(Cl)=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)C(=C(N)O)C1=O
Form
Powder
Loss on Drying
2.0% max.
Appearance (Color)
Yellow
Assay (unspecified)
Potency: 900?g/mg min.
pH
2.3 to 3.3 (1% w/v in water)

Beschreibung

Chlortetracycline was used in fluorescence assays to detect the Ca+2 influx required for the completion of acrosome reaction in human spermatozoa.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlortetracycline was used in fluorescence assays to detect the Ca+2 influx required for the completion of acrosome reaction in human spermatozoa.

Solubility
Soluble in water

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P312 - Call a POISON CENTER or doctor if you feel unwell