Thermo Scientific Chemicals

Potassium iodide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010978.22
100 g, Each
Thermo Scientific Chemicals

Potassium iodide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010978.22
100 g, Each
Quantity
Catalog number: 010978.22
also known as 010978-22
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
7681-11-0
IUPAC Name
potassium iodide
Molecular Formula
IK
InChI Key
NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES
[K+].[I-]
Total Metal Impurities
0.005% max.

Description

Potassium iodide is used with silver nitrate to make silver iodide an important chemical in film photography. Medicinally it is used for thyroid blockade, thyroid storm and also as an expectorant. It is a component in some hair treatment chemicals. It is also used as a fluorescence quenching agent in biomedical research, an application that takes advantage of collisional quenching of fluorescent substances by the iodide ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Specific target organ toxicity - (repeated exposure)
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H372 - Causes damage to organs through prolonged or repeated exposure

    Precautionary Statements

    P260 - Do not breathe dust/fume/gas/mist/vapors/spray

    P314 - Get medical advice/attention if you feel unwell