Platinum foil, 0.1mm (0.004in) thick, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum foil, 0.1mm (0.004in) thick, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number 011509.FI
also known as 011509-FI
Price (EUR)/ Each
-
Quantity:
50 x 50 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Comment0.09 - 0.11mm thick
Total Metal Impurities0.01% max.
FormFoil
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only