Thermo Scientific Chemicals

Sodium molybdenum oxide dihydrate, ACS, 99.5-103.0%, Thermo Scientific Chemicals

Catalog number: 012214.36
500 g, Each
Thermo Scientific Chemicals

Sodium molybdenum oxide dihydrate, ACS, 99.5-103.0%, Thermo Scientific Chemicals

Catalog number: 012214.36
500 g, Each
Quantity
Catalog number: 012214.36
also known as 012214-36
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
10102-40-6
IUPAC Name
disodium dioxomolybdenumbis(olate)
Molecular Formula
MoNa2O4
InChI Key
TVXXNOYZHKPKGW-UHFFFAOYSA-N
SMILES
[Na+].[Na+].[O-][Mo]([O-])(=O)=O
Impurity content
Fe 0.001%
Insoluble material
0.005%
Impurity content
NH₄ 0.001%
Impurity content
PO₄ 5ppm
Impurity content
SO₄ 0.015%

Description

It is widely used as a fertilizer in agriculture, and a corrosion inhibitor being a non-oxidizing anodic inhibitor. It reduces galvanic corrosion potential, useful in industrial waste water treatment. It is recommended for treatment of whiptail in broccoli and cauliflower. It is a precursor for molybdenum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text