Thermo Scientific Chemicals

Nickel Chromium wire, 0.143mm (0.0056 in.) dia., Thermo Scientific Chemicals

Catalog number: 013966.H9
500 m, Each
Thermo Scientific Chemicals

Nickel Chromium wire, 0.143mm (0.0056 in.) dia., Thermo Scientific Chemicals

Catalog number: 013966.H9
500 m, Each
Quantity
Catalog number: 013966.H9
also known as 013966-H9
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
11106-97-1
IUPAC Name
chromium nickel
Molecular Formula
CrNi
InChI Key
VNNRSPGTAMTISX-UHFFFAOYSA-N
SMILES
[Cr].[Ni]
Comment
0.143 mm diameter
Comment
Ni 80% min

Description

High resistivity material for electronics industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Sensitization
    Category 1
    Carcinogenicity
    Category 2
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H317 - May cause an allergic skin reaction

    H351 - Suspected of causing cancer

    Precautionary Statements

    P201 - Obtain special instructions before use

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P308 + P313 - IF exposed or concerned: Get medical advice/attention