Thermo Scientific Chemicals

Lead(II) zirconium oxide, 99.7% (metals basis excluding Hf), Hf 1% max, Thermo Scientific Chemicals

Catalog number: 014682.36
500 g, Each
Thermo Scientific Chemicals

Lead(II) zirconium oxide, 99.7% (metals basis excluding Hf), Hf 1% max, Thermo Scientific Chemicals

Catalog number: 014682.36
500 g, Each
Quantity
Catalog number: 014682.36
also known as 014682-36
Price (EUR)

Chemical Identifiers

CAS
12060-01-4
IUPAC Name
λ²-lead(2+) oxozirconiumbis(olate)
Molecular Formula
O3PbZr
InChI Key
RSCSNKLWUCUPDE-UHFFFAOYSA-N
SMILES
[Pb++].[O-][Zr]([O-])=O
Total Metal Impurities excluding Hf
0.3% max.
Impurity content
Hf 1% max

Description

It is practically applied in the area of electroceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Reproductive Toxicity
    Category 1A
    Specific target organ toxicity - (repeated exposure)
    Category 2
    Acute aquatic toxicity
    Category 1
    Chronic aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 + H332 - Harmful if swallowed or if inhaled

    H360Df - May damage the unborn child. Suspected of damaging fertility

    H373 - May cause damage to organs through prolonged or repeated exposure

    H410 - Very toxic to aquatic life with long lasting effects

    Precautionary Statements

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    Additional EU labelling

    Restricted to professional users