1x1 image pixel for data collection
Eutech™ KCl Calibration Solution
Product Image
Thermo Scientific™
Eutech™ KCl Calibration Solution
For conductivity calibration and standards, choose from our available Eutech™'s Conductivity Standards, Calibrations, and Solutions
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityConductivity
01X60920160 mL84 μS/cm
ECCON84BT480 mL84 μS/cm
ECCON100BT480 mL100 μS/cm
ECCON500BT480 mL500 μS/cm
ECCON1413BT480 mL1413 μS/cm
01X60830160 mL1413 μS/cm
ECCON2764BT480 mL2764 μS/cm
ECCON1288BT480 mL12.88 mS/cm
01X60840160 mL12.88 mS/cm
ECCON1118BT480 mL111.8 mS/cm
Catalog number 01X609201
Price (EUR)
10,00
Each
Add to cart
Quantity:
60 mL
Conductivity:
84 μS/cm
Price (EUR)
10,00
Each
Add to cart
Eutech™ KCl Calibration Solutions are used for calibrating conductivity probes for accurate measurement. Get quick and accurate conductivity measurements using Eutech™ conductivity meters and probes in the field, laboratories, and schools.
Features:
  • Packed in 60mL or 480 ml durable plastic bottles
  • Economical and convenient to use
Specifications
Conductivity84 μS/cm
Description84 μS/cm KCl Calibration Solution
For Use With (Equipment)Conductivity Meters and Electrodes
Quantity60 mL
Solution TypeCalibration Solution
TypeCalibration Solution
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link