Gadolinium(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gadolinium(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number 035737.09
also known as 035737-09
Price (EUR)/ Each
198,47
Online exclusive
223.00 
Save 24,53 (11%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (EUR)/ Each
198,47
Online exclusive
223.00 
Save 24,53 (11%)
Add to cart
Gadolinium(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
Catalog number035737.09
Price (EUR)/ Each
198,47
Online exclusive
223.00 
Save 24,53 (11%)
-
Add to cart
Chemical Identifiers
CAS13818-75-2
IUPAC Namegadolinium(3+) tribromide
Molecular FormulaBr3Gd
InChI KeyKGOKDPWKDBWITQ-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[Gd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only