Thermo Scientific Chemicals

Sodium oxalate, ACS, 99.5+%, Thermo Scientific Chemicals

Catalog number: 041759.18
50 g, Each
Thermo Scientific Chemicals

Sodium oxalate, ACS, 99.5+%, Thermo Scientific Chemicals

Catalog number: 041759.18
50 g, Each
Quantity
Catalog number: 041759.18
also known as 041759-18
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
62-76-0
IUPAC Name
disodium oxalate
Molecular Formula
C2Na2O4
InChI Key
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C(=O)C([O-])=O
Comment
Neutrality P.T.
Impurity content
Cl 0.002%
Impurity content
K 0.005%
Insoluble material
0.005%
Loss on Drying
at 105? 0.01%

Description

Sodium oxalate is used as a reducing agent, as a primary standard for standardizing potassium permanganate solutions and as a neutralizer for exothermic acidic reactions. It finds application in metal cleaning preparations, metal extraction and separation, blueprint coatings, electroplating baths, special cement manufacturing, plant nutrition as well as in textile and pharmaceutical industries. It plays an important role to remove calcium ions from blood plasma and prevents blood clotting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H312 - Harmful if swallowed or in contact with skin

    Precautionary Statements

    P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water