Calcium 3-hydroxy-3-methylbutyrate hydrate, 97+%, Thermo Scientific Chemicals
Calcium 3-hydroxy-3-methylbutyrate hydrate, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium 3-hydroxy-3-methylbutyrate hydrate, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Catalog number 042721.30
also known as 042721-30
Price (EUR)/ Each
315,06
Online exclusive
354,00 
Save 38,94 (11%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (EUR)/ Each
315,06
Online exclusive
354,00 
Save 38,94 (11%)
Add to cart
Calcium 3-hydroxy-3-methylbutyrate hydrate, 97+%, Thermo Scientific Chemicals
Catalog number042721.30
Price (EUR)/ Each
315,06
Online exclusive
354,00 
Save 38,94 (11%)
-
Add to cart
Chemical Identifiers
IUPAC Namecalcium bis(3-hydroxy-3-methylbutanoate)
Molecular FormulaC10H18CaO6
InChI KeyWLJUMPWVUPNXMF-UHFFFAOYSA-L
SMILES[Ca++].CC(C)(O)CC([O-])=O.CC(C)(O)CC([O-])=O
Molecular Weight (g/mol)274.33
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Titration)>/= 97.0%
Formpowder
Calcium 3-hydroxy-3-methylbutyrate hydrate is a nutritional supplement, which represents a calcium salt of beta-hydroxy beta-methylbutyrate (HMB). The latter is a leucine metabolite produced in the body through oxidation of the ketoacid of leucine (ketoisocaproate, KIC). Because leucine plays an important regulatory role in protein metabolism and due to the conversion of leucine to KIC, several researchers have presumed that KIC is the active ingredient responsible for most of the beneficial effects of leucine on protein metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only