Glassy carbon rod, 3mm (0.1in) dia, type 1, Thermo Scientific Chemicals
Glassy carbon rod, 3mm (0.1in) dia, type 1, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glassy carbon rod, 3mm (0.1in) dia, type 1, Thermo Scientific Chemicals

Used as an electrode material in electrochemistry. It is also employed as an electrode material for the fabrication of sensors. It serves as an ideal material for the vacuum technology. It is also used in casting, smelting of metal, hard alloys and high temperature crucibles.
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mm
100 mm
200 mm
Catalog number 042822.DJ
also known as 042822-DJ
Price (EUR)/ Each
202,50
Online exclusive
225.00 
Save 22,50 (10%)
-
Add to cart
Quantity:
50 mm
Request bulk or custom format
Price (EUR)/ Each
202,50
Online exclusive
225.00 
Save 22,50 (10%)
Add to cart
Glassy carbon rod, 3mm (0.1in) dia, type 1, Thermo Scientific Chemicals
Catalog number042822.DJ
Price (EUR)/ Each
202,50
Online exclusive
225.00 
Save 22,50 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
FormRod, 3mm (0.1in) diameter (Glassy Carbon type-1, maximimum service temperature 1000?C).
Assay from Supplier's CofA>99.9%
Glassy carbon rod is used as an electrode material in electrochemistry. It is also employed as an electrode material for the fabrication of sensors. It serves as an ideal material for the vacuum technology. It is also used in casting, smelting of metal, hard alloys and high temperature crucibles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only