Graphite foil, 0.5mm (0.02in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Graphite foil, 0.5mm (0.02in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
15 x 250 cm
15 x 15 cm
15 x 60 cm
Catalog number 042953.VU
Price (EUR)/ Each
-
Quantity:
15 x 250 cm
Request bulk or custom format
Chemical Identifiers
CAS7782-42-5
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace Metals0.2% trace metal impurities
Appearance (Color)Dark grey to black
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only