Platinum Iridium wire, 1.0mm (0.04in) dia, hard, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum Iridium wire, 1.0mm (0.04in) dia, hard, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 cm
10 cm
50 cm
Catalog number 044732.BQ
also known as 044732-BQ
Price (EUR)/ Each
-
Quantity:
5 cm
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
FormWire, 1.0mm (0.04in) diameter, hardened.
Appearance (Color)Silver gray
Assay from Supplier's CofAPt:Ir; 80:20 wt%
Platinum Iridium wire, 1.0mm (0.04in) dia, hard is used in non-standard thermocouples, electrical contacts and as electrodes in medical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only