Thermo Scientific Chemicals

Barium dodecairon nonadecaoxide, Thermo Scientific Chemicals

Catalog number: 045552.22
100 g, Each
Thermo Scientific Chemicals

Barium dodecairon nonadecaoxide, Thermo Scientific Chemicals

Catalog number: 045552.22
100 g, Each
Quantity
Catalog number: 045552.22
also known as 045552-22
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
12047-11-9
Appearance (Color)
Dark red-black
Assay from Supplier's CofA
≥94%
Form
Powder

Description

Barium dodecairon nonadecaoxide is used in subwoofer magnets to provide power. It is also used in new high tech magnetic tapes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H332 - Harmful if swallowed or if inhaled

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell