Dichloro[bis(dicyclohexylphosphino)propane]palladium(II), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloro[bis(dicyclohexylphosphino)propane]palladium(II), Thermo Scientific Chemicals
 
Have Questions?
Quantity:
1 g
Catalog number 046780.03
also known as 046780-03
Price (EUR)/ Each
333,00
Online exclusive
370.00 
Save 37,00 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)/ Each
333,00
Online exclusive
370.00 
Save 37,00 (10%)
Add to cart
Dichloro[bis(dicyclohexylphosphino)propane]palladium(II), Thermo Scientific Chemicals
Catalog number046780.03
Price (EUR)/ Each
333,00
Online exclusive
370.00 
Save 37,00 (10%)
-
Add to cart
Chemical Identifiers
CAS1041005-52-0
IUPAC Namedichloropalladium; dicyclohexyl[3-(dicyclohexylphosphaniumyl)propyl]phosphanium
Molecular FormulaC27H52Cl2P2Pd
InChI KeyJLEVAMQVDDHWOI-UHFFFAOYSA-N
SMILESCl[Pd]Cl.C(C[PH+](C1CCCCC1)C1CCCCC1)C[PH+](C1CCCCC1)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)C 51.8-53.8%
Dichloro[bis(dicyclohexylphosphino)propane]palladium(II) is used as a catalyst in coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only