Aminopyrazine, 99+%, Thermo Scientific Chemicals
Aminopyrazine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aminopyrazine, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 104490500
Price (EUR)/ Each
176,80
Online exclusive
208,00 
Save 31,20 (15%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)/ Each
176,80
Online exclusive
208,00 
Save 31,20 (15%)
Add to cart
Aminopyrazine, 99+%, Thermo Scientific Chemicals
Catalog number104490500
Price (EUR)/ Each
176,80
Online exclusive
208,00 
Save 31,20 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namepyrazin-2-amine
Molecular FormulaC4H5N3
InChI KeyXFTQRUTUGRCSGO-UHFFFAOYSA-N
SMILESNC1=CN=CC=N1
Molecular Weight (g/mol)95.11
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to beige
Infrared spectrumConforms
Titration with HClO4>=99.0 %
Appearance (Form)Crystalline powder
Melting point117°C to 120°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only