tert-Butylamine, 99%
tert-Butylamine, 99%
tert-Butylamine, 99%
tert-Butylamine, 99%
Thermo Scientific Chemicals

tert-Butylamine, 99%

CAS: 75-64-9 | C4H11N | 73.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
10 L
5 mL
2.5 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 107820010
Price (EUR)
36,55
Online Exclusive
44,10
Save 7,55 (17%)
Each
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
36,55
Online Exclusive
44,10
Save 7,55 (17%)
Each
Add to cart
Chemical Identifiers
CAS75-64-9
IUPAC Name2-methylpropan-2-amine
Molecular FormulaC4H11N
InChI KeyYBRBMKDOPFTVDT-UHFFFAOYSA-N
SMILESCC(C)(C)N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Clear liquid
Color scale=<10 APHA
Water=<0.1 %
Refractive index1.3770 to 1.3790 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link