1x1 image pixel for data collection
2-Cyclopentenone, 98%
2-Cyclopentenone, 98%
2-Cyclopentenone, 98%
2-Cyclopentenone, 98%
Thermo Scientific Chemicals

2-Cyclopentenone, 98%

CAS: 930-30-3 | C5H6O | 82.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 111570250
Price (EUR)
153,85
Online Exclusive
186,00
Save 32,15 (17%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
153,85
Online Exclusive
186,00
Save 32,15 (17%)
Each
Add to cart
Chemical Identifiers
CAS930-30-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4795 to 1.4835 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0455542Certificate of AnalysisMay 17, 2025111571000, 111570250, 111570050
A0445830Certificate of AnalysisDec 14, 2023111571000, 111570250, 111570050
A0455542Certificate of AnalysisAug 25, 2023111571000, 111570250, 111570050
A0405222Certificate of AnalysisDec 12, 2022111571000, 111570250, 111570050
A0433088Certificate of AnalysisSep 07, 2022111571000, 111570250, 111570050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Flammable liquids
Category 3
Acute oral toxicity
Category 4
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H302 - Harmful if swallowed

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

Physical Hazards

H226 - Flammable liquid and vapor

Precautionary Statements

P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

Share catalog number, name or link