Sodium bisulfate monohydrate, 99%, for analysis
Sodium bisulfate monohydrate, 99%, for analysis
Sodium bisulfate monohydrate, 99%, for analysis
Sodium bisulfate monohydrate, 99%, for analysis
Thermo Scientific Chemicals

Sodium bisulfate monohydrate, 99%, for analysis

CAS: 10034-88-5 | H3NaO5S | 138.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic drum
Catalog number 125000100
Price (EUR)
313,65
キャンペーン価格
369,00
Save 55,35 (15%)
Each
-
Add to cart
Quantity:
10 kg
Packaging:
Plastic drum
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
313,65
キャンペーン価格
369,00
Save 55,35 (15%)
Each
Add to cart
Chemical Identifiers
CAS10034-88-5
IUPAC Namesodium hydrate hydrogen sulfate
Molecular FormulaH3NaO5S
InChI KeyJXHZRQHZVYDRGX-UHFFFAOYSA-M
SMILESO.[Na+].OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Adhering crystals
Titration with NaOH>=98.5 %
Chloride (Cl)=<0.0005 %
Iron (Fe)=<0.002 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0468021Certificate of AnalysisJan 31, 2025125000100, 125000025, 125005000
A0465938Certificate of AnalysisSep 10, 2024125000100, 12500062, 12500096, 125000025, 125005000
A0451501Certificate of AnalysisDec 25, 2023T003112500CS, 125000100, 12500062, 12500096, 125000025, T003112500, 125005000
A0456736Certificate of AnalysisOct 05, 2023125000100, 12500062, 12500096, 125000025, 125005000
A0456735Certificate of AnalysisOct 04, 2023125000100, 12500062, 12500096, 125000025, 125005000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Serious Eye Damage/Eye Irritation
Category 1
Label Elements
Signal Word

Danger

Hazard Statements

H318 - Causes serious eye damage

Precautionary Statements

P280 - Wear eye protection/ face protection

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

Share catalog number, name or link

1x1 image pixel for data collection