Penicillin-Streptomycin-Neomycin (PSN) Antibiotic Mixture
Product Image

Penicillin-Streptomycin-Neomycin (PSN) Antibiotic Mixture

Have Questions?
Catalog number 15640071
Price (EUR)
786,00
1 each
-
Add to cart
Price (EUR)
786,00
1 each
Add to cart
Ask our AI about this Product
The antibiotics Penicillin, Streptomycin, and Neomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria. Penicillin was originally purified from the fungus Penicillium and acts by interfering directly with the turnover of the bacteria cell wall and indirectly by triggering the release of enzymes that further alter the cell wall. Streptomycin was originally purified from Streptomyces griseus and Neomycin from Streptomyces fradiae. Streptomycin and Neomycin act by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria. Life Technologies offers a variety of antibiotics and antimycotics for cell culture applications.

This solution contains 5 mg of penicillin, 5 mg of streptomycin, and 10 mg of neomycin per ml in 0.85% saline.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
Specifications
Unit Size1 each
Contents & Storage
Storage conditions: -5 to -20°C
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Share catalog number, name or link