Pyrimidine, 99%
Pyrimidine, 99%
Pyrimidine, 99%
Thermo Scientific Chemicals

Pyrimidine, 99%

CAS: 289-95-2 | C4H4N2 | 80.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 157700250
Price (EUR)
223,55
Online Exclusive
263,00
Save 39,45 (15%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
223,55
Online Exclusive
263,00
Save 39,45 (15%)
Each
Add to cart
Chemical Identifiers
CAS289-95-2
IUPAC Namepyrimidine
Molecular FormulaC4H4N2
InChI KeyCZPWVGJYEJSRLH-UHFFFAOYSA-N
SMILESC1=CN=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.5010 to 1.5050 (20°C, 589 nm)
Appearance (Color)Clear colorless to orange
Appearance (Form)Liquid or solid
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0444418Certificate of AnalysisJan 31, 2025157700050, 157700250
A0448797Certificate of AnalysisJan 06, 2023157700050, 157700250
A0444418Certificate of AnalysisJul 25, 2022157700050, 157700250
A0405384Certificate of AnalysisAug 09, 2021157700050, 157700250
4 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Flammable liquids
Category 3
Label Elements
Signal Word

Warning

Physical Hazards

H226 - Flammable liquid and vapor

Precautionary Statements

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

Share catalog number, name or link

1x1 image pixel for data collection