2-Cyclohepten-1-one, 90%, Thermo Scientific Chemicals
2-Cyclohepten-1-one, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Cyclohepten-1-one, 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 171070010
Price (EUR)/ Each
72,71
Online exclusive
81.70 
Save 8,99 (11%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)/ Each
72,71
Online exclusive
81.70 
Save 8,99 (11%)
Add to cart
2-Cyclohepten-1-one, 90%, Thermo Scientific Chemicals
Catalog number171070010
Price (EUR)/ Each
72,71
Online exclusive
81.70 
Save 8,99 (11%)
-
Add to cart
Chemical Identifiers
CAS1121-66-0
IUPAC Namecyclohept-2-en-1-one
Molecular FormulaC7H10O
InChI KeyWZCRDVTWUYLPTR-UHFFFAOYSA-N
SMILESO=C1CCCCC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=89.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only