Dichloromethane-d2, for NMR, 99.5 atom % D, Thermo Scientific Chemicals
Dichloromethane-d2, for NMR, 99.5 atom % D, Thermo Scientific Chemicals
Thermo Scientific™

Dichloromethane-d2, for NMR, 99.5 atom % D, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
5 mL
25 mL
50 mL
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 176110100
Price (EUR)/ Each
-
Quantity:
10 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namedichloro(²H₂)methane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-DICFDUPASA-N
SMILES[2H]C([2H])(Cl)Cl
Molecular Weight (g/mol)86.94
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.04 % (Coulometric)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
D-Enrichment>=99.4 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only