Naphazoline hydrochloride, 99%
Naphazoline hydrochloride, 99%
Naphazoline hydrochloride, 99%
Naphazoline hydrochloride, 99%
Thermo Scientific Chemicals

Naphazoline hydrochloride, 99%

CAS: 550-99-2 | C14H14N2·ClH | 246.74 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 179661000
Price (EUR)
126,65
Online Exclusive
153,00
Save 26,35 (17%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
126,65
Online Exclusive
153,00
Save 26,35 (17%)
Each
Add to cart
Chemical Identifiers
CAS550-99-2
IUPAC Namehydrogen 2-[(naphthalen-1-yl)methyl]-4,5-dihydro-1H-imidazole chloride
Molecular FormulaC14H15ClN2
InChI KeyDJDFFEBSKJCGHC-UHFFFAOYSA-N
SMILES[H+].[Cl-].C(C1=NCCN1)C1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystalline powder or crystals
Appearance (Color)White to light beige
Infrared spectrumConforms
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 2
Label Elements
Signal Word

Danger

Hazard Statements

H300 - Fatal if swallowed

Precautionary Statements

P201 - Obtain special instructions before use

P270 - Do not eat, drink or smoke when using this product

P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

P330 - Rinse mouth

Share catalog number, name or link

1x1 image pixel for data collection