1x1 image pixel for data collection
Trypan Blue
Trypan Blue
Trypan Blue
Thermo Scientific Chemicals

Trypan Blue

CAS: 72-57-1 | C34H24N6Na4O14S4 | 960.79 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 189351000
Price (EUR)
101,15
Online Exclusive
122,00
Save 20,85 (17%)
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
101,15
Online Exclusive
122,00
Save 20,85 (17%)
Each
Add to cart
Chemical Identifiers
CAS72-57-1
IUPAC Nametetrasodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-8-amino-1-oxo-3,6-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Molecular FormulaC34H24N6Na4O14S4
InChI KeyWMYVHJWZUUEZNE-ARWFNKCKSA-J
SMILES[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1\C(=O)C2=C(N)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2\C(=O)C3=C(N)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Dye content>=60 %
UV spectrumComplies
Appearance (Form)Powder
Appearance (Color)Dark blue to brown-black
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link