New Fuchsin, Thermo Scientific Chemicals
New Fuchsin, Thermo Scientific Chemicals
Thermo Scientific Chemicals

New Fuchsin, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 212140250
Price (EUR)/ Each
33,48
Online exclusive
37.20 
Save 3,72 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)/ Each
33,48
Online exclusive
37.20 
Save 3,72 (10%)
Add to cart
New Fuchsin, Thermo Scientific Chemicals
Catalog number212140250
Price (EUR)/ Each
33,48
Online exclusive
37.20 
Save 3,72 (10%)
-
Add to cart
Chemical Identifiers
CAS3248-91-7
IUPAC Nametris(4-amino-3-methylphenyl)methylium chloride
Molecular FormulaC22H24ClN3
InChI KeyORDGVBRMUJCHEO-UHFFFAOYSA-M
SMILES[Cl-].CC1=CC(=CC=C1N)[C+](C1=CC=C(N)C(C)=C1)C1=CC=C(N)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Absorption Ratio P-15/P+151.1 to 1.4
UV Lambda max550 to 554 nm (in 50% ethanol)
Appearance (Form)Crystalline powder or crystals
Specific absorbance A (1%/1cm)>=1500 (at lambda max.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only