Ethanol, 99.7%, pure, anhydrous, denat. with 3% v/v diethyl ether
Ethanol, 99.7%, pure, anhydrous, denat. with 3% v/v diethyl ether
Ethanol, 99.7%, pure, anhydrous, denat. with 3% v/v diethyl ether
Ethanol, 99.7%, pure, anhydrous, denat. with 3% v/v diethyl ether
Thermo Scientific Chemicals

Ethanol, 99.7%, pure, anhydrous, denat. with 3% v/v diethyl ether

CAS: 64-17-5 | C2H6O | 46.069 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
25 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 270660025
Price (EUR)
-
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS64-17-5
IUPAC Nameethanol
Molecular FormulaC2H6O
InChI KeyLFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILESCCO
View more
SpecificationsSpecification SheetSpecification Sheet
DenaturantDenatured with 3 % v/v diethyl ether
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For Manufacturing and Laboratory Use. Not for beverage use
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0464231Certificate of AnalysisJul 04, 2024270660025, 270660010, 270660250
A0454450Certificate of AnalysisAug 25, 2023270660025, 270660010, 270660250
A0403515Certificate of AnalysisDec 15, 2022270660025, 270660010, 270660250
A0410829Certificate of AnalysisDec 13, 2022270660025, 270660010, 270660250
A0442867Certificate of AnalysisJun 14, 2022270660025, 270660010, 270660250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Flammable liquids
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Label Elements
Signal Word

Danger

Hazard Statements

H319 - Causes serious eye irritation

Physical Hazards

H225 - Highly flammable liquid and vapor

EU Specific Hazard Statements

EUH019 - May form explosive peroxides

EUH066 - Repeated exposure may cause skin dryness or cracking

Precautionary Statements

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P264 - Wash face, hands and any exposed skin thoroughly after handling

P280 - Wear eye protection/ face protection

P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P337 + P313 - If eye irritation persists: Get medical advice/attention

Share catalog number, name or link

1x1 image pixel for data collection