Kryptofix™ 222, 98%
Kryptofix™ 222, 98%
Kryptofix™ 222, 98%
Kryptofix™ 222, 98%
Thermo Scientific Chemicals

Kryptofix™ 222, 98%

CAS: 23978-09-8 | C18H36N2O6 | 376.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 291950010
Price (EUR)
136,00
Online Exclusive
160,00
Save 24,00 (15%)
Each
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
136,00
Online Exclusive
160,00
Save 24,00 (15%)
Each
Add to cart
Chemical Identifiers
CAS23978-09-8
IUPAC Name4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane
Molecular FormulaC18H36N2O6
InChI KeyAUFVJZSDSXXFOI-UHFFFAOYSA-N
SMILESC1COCCN2CCOCCOCCN(CCO1)CCOCCOCC2
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point68°C to 72°C
Appearance (Form)Crystalline powder or crystals
GC>=97.5 %
Appearance (Color)White
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473524Certificate of AnalysisJun 24, 2025291952500, 291950010, 291950050
A0443994Certificate of AnalysisJan 31, 2025291952500, 291950010, 291950050
A0462930Certificate of AnalysisJun 10, 2024291952500, 291950010, 291950050
A0402104Certificate of AnalysisDec 14, 2022291952500, 291950010, 291950050
A0393641Certificate of AnalysisDec 14, 2022291952500, 291950010, 291950050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H302 - Harmful if swallowed

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

Precautionary Statements

P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

Share catalog number, name or link

1x1 image pixel for data collection