3-Methoxyphenethylamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Methoxyphenethylamine, 98+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 302610250
Price (EUR)/ Each
202,03
Online exclusive
227.00 
Save 24,97 (11%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)/ Each
202,03
Online exclusive
227.00 
Save 24,97 (11%)
Add to cart
3-Methoxyphenethylamine, 98+%, Thermo Scientific Chemicals
Catalog number302610250
Price (EUR)/ Each
202,03
Online exclusive
227.00 
Save 24,97 (11%)
-
Add to cart
Chemical Identifiers
CAS2039-67-0
IUPAC Namehydrogen 2-(3-methoxyphenyl)ethan-1-amine chloride
Molecular FormulaC9H14ClNO
InChI KeyKDRBQDYHPKRFGX-UHFFFAOYSA-N
SMILES[H+].[Cl-].COC1=CC=CC(CCN)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.0 %
Appearance (Color)Clear colorless to yellow
Titration with HClO4>=98.0 %
Refractive index1.5375 to 1.5395 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only