Dionex™ ERS™ 500e Electrolytically Regenerated Suppressor
Dionex™ ERS™ 500e Electrolytically Regenerated Suppressor
Actual product may vary
Thermo Scientific™

Dionex™ ERS™ 500e Electrolytically Regenerated Suppressor

This next-generation suppressor delivers high-capacity, low noise, high backpressure resiliency, and fast startup to meet today's demands in ion chromatography.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
302662Dionex AERS 500 e Anion Electrolytically Regenerated Suppressor for External Water Mode (2 mm)
302661Dionex AERS 500 e Anion Electrolytically Regenerated Suppressor for External Water Mode (4 mm)
302663Dionex CERS 500 e Cation Electrolytically Regenerated Suppressor for External Water Mode (4 mm)
302664Dionex CERS 500 e Cation Electrolytically Regenerated Suppressor for External Water Mode (2 mm)
4 Options
Catalog number 302662
Price (EUR)/ Each 1
2.350,00
-
Add to cart
Description:
Dionex AERS 500 e Anion Electrolytically Regenerated Suppressor for External Water Mode (2 mm)
Price (EUR)/ Each 1
2.350,00
Add to cart
Dionex™ ERS™ 500e Electrolytically Regenerated Suppressor
Catalog number302662
Price (EUR)/ Each 1
2.350,00
-
Add to cart

With high-capacity, low noise, high backpressure resiliency, and fast startup, this new generation suppressor was designed for new demands in IC. The Thermo Scientific™ Dionex™ ERS™ 500e Electrolytically Regenerated Suppressor for external water mode applications minimizes peak dispersion and improves peak efficiency, particularly when coupled with 4μm columns. This suppressors was developed in concert with the high efficiency 4μm ion-exchange columns, which use higher flow rates to achieve faster separations while retaining optimal resolution, or equivalent flow rates to achieve higher resolutions while retaining sample throughput.

This high-capacity suppressor serves virtually all analytical scale IC applications, for both anions and cations, and is available in 2mm and 4mm formats. The Dionex ERS 500e Suppressor accommodates isocratic or gradient use with carbonate (isocratic only) hydroxide, methanesulfonic acid, or sulfuric acid eluents in both the standard bore (4 mm) and microbore (2 mm) format.

Next-Generation Suppressor—Built for New Demands in IC Applications
  • Improved compatibility with hyphenated detection techniques, particularly mass spectrometry, because of its higher backpressure tolerance (leak free up to 900 psi, operates up to 200 psi)
  • Higher temperature tolerance (up to 80°C in transit and storage, 40°C for operation)
  • Full electrolytic regeneration without reagents allowing easier recoveries after operation without current due to its planar packed-bed suppression chamber
  • Low peak dispersion preserves peak shape efficiency of 4μm columns
  • High suppression capacity produces stable peak area values immediately after restart without the need for chemical regeneration
  • Low noise makes the suppressor suitable for trace analysis
Built for External Water Mode Applications
  • Reduced regenerant chamber backpressure for easy implementation of pressurized reservoir delivery.
  • Parallel regenerant flow to maximize regenerant sweep out for borate eluents and eluents containing solvents (up to 40%).
  • Can be used in the Chemical Regenerant Mode.
Improves Sensitivity for More Reliable Quantitation

When used with the Thermo Scientific™ Dionex™ CRD 300 Carbonate Removal Device, the Dionex ERS 500e Suppressor reduces background conductivity and noise, enhances analyte response, and improves sensitivity for more reliable quantitation with carbonate eluents.

Fundamental to Enabling Reagent-Free™ IC (RFIC™) Operation
  • Full regeneration without reagents

The Dionex ERS 500e suppressor is a direct replacements for the Thermo Scientific™ Dionex™ SRS™ 300 Self-Regenerating Suppressor.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for six (6) months thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
TypeAnion Electrolytically Regenerated Suppressor
Diameter4.8 cm (1.9 in.)
Flow RateMaximum Eluent Flow Rate: 0.75 mL/min.
PressureRecommended Backpressure: 30 to 150 psi
SolventsExternal Water Mode Up to 40%<br /> Methanol, Ethanol, Acetonitrile, Isopropanol
OperationDesigned with high-capacity, low noise, high backpressure resiliency, and fast startup.
Weight (English)0.65 lb.
Weight (Metric)295 g
Width (Metric)4.5 cm
Amperage150 mA
For Use With (Application)Modes of Operation Supported: External Water Mode for Eluents Containing up to 40% Solvent; Chemical Regeneration Mode for Eluents Containing up to 100% Solvent; Recycled Eluent Mode for Aqueous Eluents Without Solvent; MPIC (Mobile Phase Ion Chromatography) Mode for Ion-Pairing Eluents
Temperature RangeOperational Temperature Range: 15°C to 40°C<br /> When installed outside the heated column enclosure, suppressors can support applications up to 60°C.<br /> Shipping Temperature Range: 0°C to 70°C<br /> Short-term only; do not store or operate suppressors above 40°C for extended lengths of time.
Void VolumeLess than 15 μL
DescriptionDionex AERS 500 e Anion Electrolytically Regenerated Suppressor for External Water Mode (2 mm)
Unit SizeEach 1