Methyl 3-(4-hydroxyphenyl)propionate, 98%, Thermo Scientific Chemicals
Methyl 3-(4-hydroxyphenyl)propionate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 3-(4-hydroxyphenyl)propionate, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
5 g
Packaging:
Glass bottle
Catalog number 369430050
Price (EUR)/ Each
45,54
Online exclusive
50.60 
Save 5,06 (10%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)/ Each
45,54
Online exclusive
50.60 
Save 5,06 (10%)
Add to cart
Methyl 3-(4-hydroxyphenyl)propionate, 98%, Thermo Scientific Chemicals
Catalog number369430050
Price (EUR)/ Each
45,54
Online exclusive
50.60 
Save 5,06 (10%)
-
Add to cart
Chemical Identifiers
CAS5597-50-2
IUPAC Namemethyl 3-(4-hydroxyphenyl)propanoate
Molecular FormulaC10H12O3
InChI KeyXRAMJHXWXCMGJM-UHFFFAOYSA-N
SMILESCOC(=O)CCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point37°C to 41°C
GC>=97.5 %
Appearance (Color)White to beige
Appearance (Form)Crystalline powder or crystals or fused mass
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only