Mexiletine hydrochloride, Thermo Scientific Chemicals
Mexiletine hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mexiletine hydrochloride, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 372911000
Price (EUR)/ Each
172,00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)/ Each
172,00
Add to cart
Mexiletine hydrochloride, Thermo Scientific Chemicals
Catalog number372911000
Price (EUR)/ Each
172,00
-
Add to cart
Chemical Identifiers
CAS5370-01-4
IUPAC Namehydrogen 1-(2,6-dimethylphenoxy)propan-2-amine chloride
Molecular FormulaC11H18ClNO
InChI KeyNFEIBWMZVIVJLQ-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(N)COC1=C(C)C=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Melting point200°C to 205°C
Loss on drying=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only