Castor oil
Castor oil
Castor oil
Thermo Scientific Chemicals

Castor oil

CAS: 8001-79-4 | C57H104O9 | 933.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
2.5 kg
Catalog number 397282500
Price (EUR)
71,23
Precio exclusivo en nuestra web
83,80
Save 12,57 (15%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (EUR)
71,23
Precio exclusivo en nuestra web
83,80
Save 12,57 (15%)
Each
Add to cart
Chemical Identifiers
CAS8001-79-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
pH6.0 to 8.0 (10 % in water)
Water=<3.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link