Sulfapyridine, 95%, Thermo Scientific Chemicals
Sulfapyridine, 95%, Thermo Scientific Chemicals
Sulfapyridine, 95%, Thermo Scientific Chemicals
Sulfapyridine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfapyridine, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
44285025025 g
Catalog number 442850250
Price (EUR)
40,46
キャンペーン価格
47,60
Save 7,14 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
40,46
キャンペーン価格
47,60
Save 7,14 (15%)
Each
Add to cart
Chemical Identifiers
IUPAC Name4-amino-N-(pyridin-2-yl)benzene-1-sulfonamide
Molecular FormulaC11H11N3O2S
InChI KeyGECHUMIMRBOMGK-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=N1
Molecular Weight (g/mol)249.29
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=94 %
Appearance (Color)Yellow
Melting point190°C to 193°C
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link