Chemical Identifiers
IUPAC Name4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Molecular FormulaC9H11F2N3O4
InChI KeySDUQYLNIPVEERB-QPPQHZFASA-N
SMILESNC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)C1(F)F
Molecular Weight (g/mol)263.20
View more