Erlotinib hydrochloride
Erlotinib hydrochloride
Thermo Scientific Chemicals

Erlotinib hydrochloride

CAS: 183319-69-9 | C22H24ClN3O4 | 429.90 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 462330010
Price (EUR)
91,80
Online Exclusive
111,00
Save 19,20 (17%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
91,80
Online Exclusive
111,00
Save 19,20 (17%)
Each
Add to cart
Chemical Identifiers
CAS183319-69-9
IUPAC NameN-(3-ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride
Molecular FormulaC22H24ClN3O4
InChI KeyGTTBEUCJPZQMDZ-UHFFFAOYSA-N
SMILESCl.COCCOC1=C(OCCOC)C=C2C(NC3=CC=CC(=C3)C#C)=NC=NC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
Solubility(1% in DMSO) Clear solution
Loss on drying=<1 %
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link