3,3-Diphenylpropionitrile, Thermo Scientific Chemicals
3,3-Diphenylpropionitrile, Thermo Scientific Chemicals
3,3-Diphenylpropionitrile, Thermo Scientific Chemicals
3,3-Diphenylpropionitrile, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3-Diphenylpropionitrile, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 466080050
Price (EUR)
46,16
Online Exclusive
55,70
Save 9,54 (17%)
Each
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
46,16
Online Exclusive
55,70
Save 9,54 (17%)
Each
Add to cart
Chemical Identifiers
CAS2286-54-6
IUPAC Name3,3-diphenylpropanenitrile
Molecular FormulaC15H13N
InChI KeyINERKLNEVAZSCI-UHFFFAOYSA-N
SMILESN#CCC(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Acute dermal toxicity
Category 4
Acute Inhalation Toxicity - Dusts and Mists
Category 4
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P312 - Call a POISON CENTER or doctor if you feel unwell

Share catalog number, name or link