Cyclobenzaprine hydrochloride
Cyclobenzaprine hydrochloride
Thermo Scientific Chemicals

Cyclobenzaprine hydrochloride

CAS: 6202-23-9 | C20H21N·HCl, C20H22ClN | 311.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 466250010
Price (EUR)
29,50
Online Exclusive
34,70
Save 5,20 (15%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
29,50
Online Exclusive
34,70
Save 5,20 (15%)
Each
Add to cart
Chemical Identifiers
CAS6202-23-9
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Infrared spectrumConforms
Loss on drying=<1 % (105°C)
Solubility(50 mg/ml in ethanol) clear colorless to light yellow
Appearance (Color)White to almost white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 3
Label Elements
Signal Word

Danger

Hazard Statements

H301 - Toxic if swallowed

Precautionary Statements

P264 - Wash face, hands and any exposed skin thoroughly after handling

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P405 - Store locked up

Share catalog number, name or link

1x1 image pixel for data collection