Zofenopril calcium
Thermo Scientific Chemicals

Zofenopril calcium

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466561000
Price (EUR)
106,25
Online Exclusive
128,00
Save 21,75 (17%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
100 mg
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
106,25
Online Exclusive
128,00
Save 21,75 (17%)
Each
Add to cart
Chemical Identifiers
CAS81938-43-4
IUPAC Namecalcium bis((2S,4S)-1-[(2S)-3-(benzoylsulfanyl)-2-methylpropanoyl]-4-(phenylsulfanyl)pyrrolidine-2-carboxylate)
Molecular FormulaC44H44CaN2O8S4
InChI KeyNSYUKKYYVFVMST-LETVYOFWSA-L
SMILES[Ca++].C[C@H](CSC(=O)C1=CC=CC=C1)C(=O)N1C[C@H](C[C@H]1C([O-])=O)SC1=CC=CC=C1.C[C@H](CSC(=O)C1=CC=CC=C1)C(=O)N1C[C@H](C[C@H]1C([O-])=O)SC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)powder
HPLC>=98.5 %
Infrared spectrumConforms
Loss on drying=<0.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0418816Certificate of AnalysisJul 07, 2025466561000, 466565000
A0418816Certificate of AnalysisJan 05, 2023466561000, 466565000
2 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute aquatic toxicity
Category 1
Chronic aquatic toxicity
Category 1
Label Elements
Signal Word

Warning

Hazard Statements

H410 - Very toxic to aquatic life with long lasting effects

Precautionary Statements

P273 - Avoid release to the environment

P391 - Collect spillage

P501 - Dispose of contents/ container to an approved waste disposal plant

Share catalog number, name or link

1x1 image pixel for data collection