Estramustine phosphate sodium
Estramustine phosphate sodium
Thermo Scientific Chemicals

Estramustine phosphate sodium

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4667400101 g
466742500250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466740010
Price (EUR)
490,45
Online Exclusive
591,00
Save 100,55 (17%)
Each
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
490,45
Online Exclusive
591,00
Save 100,55 (17%)
Each
Add to cart
Chemical Identifiers
CAS52205-73-9
IUPAC Namedisodium (1S,3aS,3bR,9bS,11aS)-7-{[bis(2-chloroethyl)carbamoyl]oxy}-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl phosphate
Molecular FormulaC23H30Cl2NNa2O6P
InChI KeyIIUMCNJTGSMNRO-VVSKJQCTSA-L
SMILES[Na+].[Na+].C[C@]12CC[C@H]3[C@@H](CCC4=CC(OC(=O)N(CCCl)CCCl)=CC=C34)[C@@H]1CC[C@@H]2OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)powder
Water=<12.5 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Carcinogenicity
Category 2
Reproductive Toxicity
Category 1B
Label Elements
Signal Word

Danger

Hazard Statements

H351 - Suspected of causing cancer

H360 - May damage fertility or the unborn child

Precautionary Statements

P201 - Obtain special instructions before use

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P308 + P313 - IF exposed or concerned: Get medical advice/attention

Additional EU labelling

Restricted to professional users

Share catalog number, name or link