Thermo Scientific Chemicals

Ibrutinib

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number 466800010
Price (EUR)/ Each
148,00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)/ Each
148,00
Add to cart
Ibrutinib
Catalog number466800010
Price (EUR)/ Each
148,00
-
Add to cart
Chemical Identifiers
CAS936563-96-1
SpecificationsSpecification SheetSpecification Sheet
Water=<1.0 %
Appearance (Form)powder
Solubility(0.5% in DMSO) Clear colorless solution
Appearance (Color)White to almost white
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only