Myclobutanil
Thermo Scientific Chemicals

Myclobutanil

CAS: 88671-89-0 | C15H17ClN4 | 288.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 467142500
Price (EUR)
65,71
Online Exclusive
77,30
Save 11,59 (15%)
Each
-
Add to cart
Quantity:
250 mg
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
65,71
Online Exclusive
77,30
Save 11,59 (15%)
Each
Add to cart
Chemical Identifiers
CAS88671-89-0
IUPAC Name2-(4-chlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]hexanenitrile
Molecular FormulaC15H17ClN4
InChI KeyHZJKXKUJVSEEFU-UHFFFAOYNA-N
SMILESCCCCC(CN1C=NC=N1)(C#N)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point69°C to 76°C
Water=<1 %
HPLC>=94 %
Appearance (Color)White to light yellow to light beige
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0419990Certificate of AnalysisFeb 10, 2023467142500, 467140010
1 result displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Serious Eye Damage/Eye Irritation
Category 2
Reproductive Toxicity
Category 2
Chronic aquatic toxicity
Category 2
Label Elements
Signal Word

Warning

Hazard Statements

H302 - Harmful if swallowed

H319 - Causes serious eye irritation

H361d - Suspected of damaging the unborn child

H411 - Toxic to aquatic life with long lasting effects

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P312 - Call a POISON CENTER or doctor if you feel unwell

Share catalog number, name or link

1x1 image pixel for data collection